missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Cytokeratin 8/18 Ab-1, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 8/18 Ab-1, Mouse Monoclonal Antibody.
134.00€ - 359.00€
Specifications
Antigen | Cytokeratin 8/18 Ab-1 |
---|---|
Clone | 5D3 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12662227
|
Epredia™
MS-743-S0 |
Concentrated | 100 μL |
134.00€
100µL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12672227
|
Epredia™
MS-743-S1 |
Concentrated | 500 μL |
245.00€
500µL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12642227
|
Epredia™
MS-743-S |
Concentrated | 1 mL |
359.00€
1mL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12632227
|
Epredia™
MS-743-R7 |
Prediluted | 7 mL |
193.00€
7mL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12652227
|
Epredia™
MS-743-RQ |
Ready-to-Use | 12 mL |
293.00€
12mL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with cytokeratin 18 (type A; acidic subfamily of low molecular weight keratins).Host Species: Mouse
Clone: 5D3
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Immunogen: Cytokeratins purified from breast cancer MCF-7 cells
Molecular Weight: 52.5 kDa (Cytokeratin 8) and 45 kDa (Cytokeratin 18)
Positive Control: Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Cytokeratin 8/18 Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Cytokeratins purified from breast cancer MCF-7 cells |
5D3 | |
Monoclonal | |
Mouse | |
Cytoskeletal Protein | |
Primary |