missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cytosporone B
GREENER_CHOICE

Product Code. 18727088
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18727088

Brand: Tocris Bioscience™ 5459/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity Nur77 agonist

Cytosporone B is a naturally occurring Nur77 (NR4A1) agonist (Kd = 8.52 x 10-7 M). Induces apoptosis in gastric cancer cells in vitro. Increases blood glucose levels in fasting C57 mice and suppresses xenograft tumor growth. Also exhibits a potent antifibrotic effect in bleomycin-induced skin fibrosis in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 3,5-dihydroxy-2-(1-oxooctyl)-benzeneacetate
CAS 321661-62-5
Quantity 10 mg
Target Nuclear Receptor Agonists
Molecular Formula C18H26O5
Formula Weight 322.4
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.