missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Alpha-Bromocamphor-8-sulfonic Acid Ammonium Salt, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | D-(+)-a-Bromocamphor-8-sulfonic Acid Ammonium Salt |
| CAS | 14575-84-9 |
| Molecular Formula | C10 H14 Br O4 S . H4 N |
| Quantity | 5 g |
| Synonym | Bicyclo[2.2.1]heptane-7-methanesulfonic acid, 2-bromo-4,7-dimethyl-3-oxo-, ammonium salt (1:1), (1S,2S,4R,7R)-, 8-Bornanesulfonic acid, 3-bromo-2-oxo-, ammonium salt, (+)- (8CI), Bicyclo[2.2.1]heptane-7-methanesulfonic acid, 3-bromo-1,7-dimethyl-2-oxo-, ammonium salt, (1R,3S,4S,7R)- (9CI), Bicyclo[2.2.1]heptane-7-methanesulfonic acid, 3-bromo-1,7-dimethyl-2-oxo-, ammonium salt, [1R-(endo,anti)]-, (1R)-(+)-3-Bromocamphor-8-sulfonic acid ammonium salt, Ammonium (+)-α-bromocamphor-π-sulfonate, Ammonium (1R)-(+)-3-bromocamphor-8-sulfonate, Ammonium [(1R)-(endo,anti)]-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptane-7-methanesulfonate, Ammonium d-α-bromocamphor-π-sulfonate, D-(+)-α-Bromocamphor-8-sulfonic Acid Ammonium Salt |
| InChI Formula | InChI=1S/C10H15BrO4S.H3N/c1-9-4-3-6(7(11)8(9)12)10(9,2)5-16(13,14)15;/h6-7H,3-5H2,1-2H3,(H,13,14,15);1H3/t6-,7+,9+,10-;/m1./s1 |
| SMILES | [NH4+].C[C@@]1(CS(=O)(=O)[O-])[C@@H]2CC[C@@]1(C)C(=O)[C@H]2Br |
| IUPAC Name | azanium;[(1S,2S,4R,7R)-2-bromo-4,7-dimethyl-3-oxo-7-bicyclo[2.2.1]heptanyl]methanesulfonate |
| Molecular Weight (g/mol) | 328.22 |
| Formula Weight | 327.014 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction