missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Fructose-1,6-bisphosphate trisodium salt
Click to view available options
Quantity:
1 g
5 g
25 g
Description
D-Fructose-1,6-bisphosphate trisodium salt is used to reduce ischemia-reperfusion injury in rats.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Fructose-1,6-bisphosphate trisodium salt is used to reduce ischemia-reperfusion injury in rats.
Solubility
Soluble in water at 50mg/ml
Notes
Protect from heat. Incompatible with heat and oxidizing agents.
D-Fructose-1,6-bisphosphate trisodium salt is used to reduce ischemia-reperfusion injury in rats.
Solubility
Soluble in water at 50mg/ml
Notes
Protect from heat. Incompatible with heat and oxidizing agents.
Specifications
Color | White |
Quantity | 1 g |
Solubility Information | Soluble in water at 50mg/ml |
Formula Weight | 406.06 |
Physical Form | Powder |
Chemical Name or Material | D-Fructose-1,6-bisphosphate trisodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'