missing translation for 'onlineSavingsMsg'
Learn More

D-Leucinol, 97%

Product Code. 11397986
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11397986

Brand: Thermo Scientific Alfa Aesar L13243.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 53448-09-2 | C6H16NO | 118.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53448-09-2
Molecular Formula C6H16NO
Molecular Weight (g/mol) 118.20
MDL Number MFCD00004734
InChI Key VPSSPAXIFBTOHY-ZCFIWIBFSA-O
Synonym d-leucinol, r-2-amino-4-methylpentan-1-ol, 2r-2-amino-4-methylpentan-1-ol, d---leucinol, chembl71171, r---leucinol, 1-pentanol, 2-amino-4-methyl-, 2r, 2-amino-4-methyl-pentan-1-ol, 2-amino-4-methyl-1-pentanol #
PubChem CID 2724002
IUPAC Name (2R)-2-amino-4-methylpentan-1-ol
SMILES CC(C)C[C@@H]([NH3+])CO

Specifications

Refractive Index 1.4496
Density 0.917
Boiling Point 198°C to 200°C (690mmHg)
Quantity 5 g
Flash Point 90°C (194°F)
Optical Rotation −3.2° (c=9 in Ethanol)
Sensitivity Air Sensitive
Beilstein 1719241
Formula Weight 117.19
Percent Purity 97%
Chemical Name or Material D-Leucinol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.