Learn More
Thermo Scientific Chemicals D(+)-Melezitose monohydrate, 99+%
Catalog Number p-155559
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
10030-67-8
C18H36O18
540.468
MFCD00149448
LNVIPYYEBMNJIL-ZWELICPFSA-N
melezitose dihydrate, d-melezitosedihydrate, unii-d12c11k0m0, d-melezitose dihydrate, melezitose dihydrate mi, melezitose dihydrate, +, alpha-d-glucopyranoside, o-alpha-d-glucopyranosyl-1->3-beta-d-fructofuranosyl, dihydrate, o-alpha-d-glucopyranosyl-1->3-beta-d-fructofuranosyl-alpha-d-glucopyranoside dihydrate, alpha-d-glucopyranosyl-1->3-beta-d-fructofuranosyl alpha-d-glucopyranoside-water 1/2, 2r,3r,4s,5s,6r-2-2s,3s,4r,5r-4-hydroxy-2,5-bis hydroxymethyl-2-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxolan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol dihydrate
67660249
(2R,3R,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;dihydrate
C(C1C(C(C(C(O1)OC2C(C(OC2(CO)OC3C(C(C(C(O3)CO)O)O)O)CO)O)O)O)O)O.O.O
10030-67-8
C18H36O18
540.468
MFCD00149448
LNVIPYYEBMNJIL-ZWELICPFSA-N
melezitose dihydrate, d-melezitosedihydrate, unii-d12c11k0m0, d-melezitose dihydrate, melezitose dihydrate mi, melezitose dihydrate, +, alpha-d-glucopyranoside, o-alpha-d-glucopyranosyl-1->3-beta-d-fructofuranosyl, dihydrate, o-alpha-d-glucopyranosyl-1->3-beta-d-fructofuranosyl-alpha-d-glucopyranoside dihydrate, alpha-d-glucopyranosyl-1->3-beta-d-fructofuranosyl alpha-d-glucopyranoside-water 1/2, 2r,3r,4s,5s,6r-2-2s,3s,4r,5r-4-hydroxy-2,5-bis hydroxymethyl-2-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxolan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol dihydrate
67660249
(2R,3R,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;dihydrate
C(C1C(C(C(C(O1)OC2C(C(OC2(CO)OC3C(C(C(C(O3)CO)O)O)O)CO)O)O)O)O)O.O.O
D(+)-Melezitose monohydrate, CAS # 597-12-6, is the D-form hydrate of melezitose, the trisaccharide.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Sap-feeding insects such as ants and aphids produce melezitose to maintain osmotic balance
• Melezitose is produced by some plants as an insect attractant and acts as a metabolite
Applications
• Melezitose can be hydrolyzed to glucose and turanose through alpha-glucosidase activity and can be used for the study of enzyme activity
• Melezitose can be used to distinguish certain strains of bacteria; characterization of phenotype based on the presence or absence of certain sugars assists identification
TRUSTED_SUSTAINABILITY
Melting Point 160.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Beilstein 31, 466
Merck Index 14, 5819
Specific Rotation Condition +88° (20°C c=4,H2O)
Specific Rotation + 88.00
Formula Weight 522.45
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Melezitose monohydrate, 99+%