missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol-2,3,5-trisphosphate ammonium salt, 98%, Thermo Scientific™

Catalog Number 15410957
Click to view available options
Quantity:
0.1 mg
0.5 mg
This item is not returnable. View return policy
MFCD11114547
Ins(2,3,5)-P3 ammonium salt; 2,3,5-IP3 ammonium salt
This item is not returnable. View return policy
MFCD11114547
Ins(2,3,5)-P3 ammonium salt; 2,3,5-IP3 ammonium salt

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 0.1 mg
Formula Weight 471.19
Percent Purity 98%
Physical Form Lyophilized Powder
Chemical Name or Material D-myo-Inositol-2,3,5-trisphosphate ammonium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'