missing translation for 'onlineSavingsMsg'
Learn More

D-Serine, TRC

Product Code. 30427378
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30427378 50 g 50g
30426232 5 g -
2 options
This item is not returnable. View return policy

Product Code. 30427378

Brand: Toronto Research Chemicals TRCS27097550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312-84-5
Formula Weight 105.0426
InChI Formula InChI=1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m1/s1
IUPAC Name (2R)-2-amino-3-hydroxypropanoic acid
Molecular Formula C3 H7 N O3
Molecular Weight (g/mol) 105.09
SMILES N[C@H](CO)C(=O)O
Synonym D-Serine, Serine, D- (8CI), (R)-2-Amino-3-hydroxypropanoic acid, (R)-Serine, NSC 77689
Chemical Name or Material D-Serine

Specifications

Quantity 50 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.