missing translation for 'onlineSavingsMsg'
Learn More

D-Threonine, TRC

Product Code. 30454753
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
30454753 25 g 25g
30454966 5 g 5g
30453346 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 30454753

Brand: Toronto Research Chemicals TRCT40552525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-20-2
Formula Weight 119.0582
InChI Formula InChI=1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m0/s1
IUPAC Name (2R,3S)-2-amino-3-hydroxybutanoic acid
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
SMILES [C@@H]([C@H](C)O)(C(O)=O)N
Synonym (2R,3S)-2-Amino-3-hydroxybutanoic Acid, (2R,3S)-3-Hydroxy-2-aminobutanoic Acid, (R)-Threonine, NSC 46702
Chemical Name or Material D-Threonine

Specifications

Quantity 25 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.