Learn More
Abeba™ Dark Blue Velours ESD Anatom Safety Shoes
Product Code. 15240925
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
Unit Size:
1 pair
Each
Product Code. 15240925
Brand: Abeba™ 32147/47
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S1 SRC
  • Velours with breathable textile inlays
  • Dark blue
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Suitable for heel and foot lift
  • Scratch-resistant coating in heel area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color Blue
Material Velours
Size 47
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32147
Toe Material Steel