Learn More
Abeba™ Dark Blue Velours ESD Anatom Safety Shoes
Product Code. 15528975
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
Unit Size:
1 pair
Each
Product Code. 15528975
Brand: Abeba™ 32147/45
Only null left
Add to basket
Add to basket
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S1 SRC
  • Velours with breathable textile inlays
  • Dark blue
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Suitable for heel and foot lift
  • Scratch-resistant coating in heel area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color Blue
Material Velours
Size 45
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32147
Toe Material Steel