missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Darunavir

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Darunavir is a highly potent HIV protease inhibitor (IC50 = 3 - 6 nM, depending on laboratory HIV-1 strain). Displays activity in cells infected with clinical HIV-1 strains shown to have resistance to other protease inhibitors.
Tocris products are for biomedical research use only. They are not intended for human or veterinary use.
Specifications
Specifications
| Chemical Name or Material | (3 R,3 AS,6 aR)-Hexahydrofuro[2,3-b]furan-3-yl N-[(1 S,2 R)-3-[[(4-aminophenyl)sulfonyl](2-methylpropyl)amino]-2-hydroxy-1-(phenylmethyl)propyl]carbamate |
| CAS | 206361-99-1 |
| Quantity | 50 mg |
| Target | HIV Protease Inhibitors |
| Molecular Formula | C27H37N3O7S |
| Formula Weight | 547.66 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction