Learn More
Fisherbrand™ Data Outputs for Ultra-Low Temperature Freezers
Integrate Fisherbrand™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
RS-232:- RS-232 and 4-40mA analog outputs are standard on Thermo Scientific™ Forma™ 8600 Series freezers. These additional options are designed to meet specific needs.
- RS-232 allows connection to a computer, recorder, or independent monitoring device RS-485 allows direct connection to the Model 1535 Monitor/Alarm System
Note:
RS-232 is not compatible with the Forma Model 1535 Monitor/Alarm System.
Specifications
RS-485, 0-5V (Factory Installed Only) | |
Refrigerator/Freezer Electronics | |
Forma™ 8600 | |
0/5 V |