Learn More
Emma Safety Footwear Dave (Ruffneck) Safety Shoes
Product Code. 15949071
Click to view available options
Size:
39
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Product Code. 15949071
Brand: Emma Safety Footwear 91454645
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Looks and feels like a classic sneaker. Highly supportive reinforced ruff neck heel together with the shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • S3-Safety shoe with sole plate for wet work
  • Nubuck / Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • ESD certified
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Brown
Design Dave (Ruffneck)
Liner Material Hydro-Tec™ Sanitized Silver
Material Nubuck, Oiled Nubuck
Size 45
Toe Material Steel