Learn More
Emma Safety Footwear Dave (Ruffneck) Safety Shoes
Looks and feels like a classic sneaker. Highly supportive reinforced ruff neck heel together with the shock absorbing sole keeps ankles, shins and joints free from stress and injury.
Brand:  Emma Safety Footwear 914546-46
Additional Details : Weight : 1.74000kg
 View more versions of this product
Product Code. 15959071
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Nubuck / Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • ESD certified
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Brown
Hydro-Tec™ Sanitized Silver
46
Laces
Dave (Ruffneck)
Nubuck, Oiled Nubuck
Steel