Learn More
Emma Safety Footwear Daytona Safety Shoes
Ideal for warm, enclosed environments. Features Emma's unique Hydro-Tec™ Sanitized Silver technology, removing moisture and perspiration instantly. Synthetic, reinforced toes and super swift Velcro fasteners make the Daytona breathable, wearable and incredibly comfortable.
Brand: Emma Safety Footwear MM78654046
Additional Details : Weight : 1.64300kg
Description
- S1-Safety shoe for dry working conditions
- Nubuck construction
- GripForce™ Easy Twist Outsole
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Velcro closure
- Sanitized
Specifications
EN ISO 20345:2011 | |
Black, Gray | |
Hydro-Tec™ Sanitized Silver | |
46 |
Velcro | |
Daytona | |
Nubuck | |
Steel |