missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DBC1 (Human) Recombinant Protein
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
- Sequence: MNWRFVGLLYFLFIWGRISVQPSHQEPAGTDQHVSKEFDWLISDRGPFHHSRSYLSFVERHRQGFTTRYKIYREFARWKVRNTAIERRDLVRHPVPLMPEFQRSIRLLGRRPTTQQFIDTIIKKYGTHLLISATLGGEEALTMYMDKSRLDRKSGNATQSVEALHQLASSYFVDRDGTMRRLHEIQISTGAIKVTETRTGPLGCNSYDNLDSVSSVLLQSTESKLHLQGLQIIFPQYLQEKFVQSALSYIMCNGEGEYLCQNSQCRCQCAEEFPQCNCPITDIQIMEYTLANMAKSWAEAYKDLENSGRESHSVPLHEWP
Specifications
Specifications
| Accession Number | AAH21560 |
| Gene ID (Entrez) | 1620 |
| Name | deleted in bladder cancer 1 |
| Preparation Method | Wheat germ expression system |
| Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | DBCCR1, FAM5A |
| Gene Symbol | DBC1 |
| Species | Wheat Germ (in vitro) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction