missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DBCO-NHS

Description
DBCO-NHS is an amine reactive and azide reactive building block. It can be used for copper-free click cycloaddition reactions.
Specifications
Specifications
| Chemical Name or Material | 2,5-Dioxo-1-pyrrolidinyl-11,12-didehydro-γ-oxodibenz[b,f]azocine-5(6 H)-butanoate |
| CAS | 1353016-71-3 |
| Quantity | 25 mg |
| Target | Buffers, Chelators and Reagents |
| Molecular Formula | C23H18N2O5 |
| Purity | 0.95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?