Learn More
DCUN1D3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560396
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: IALWKLVFTQ NNPPVLDQWL NFLTENPSGI KGISRDTWNM FLNFTQVIGP DLSNYSEDEA WPSLFDTFVE WEMERRKR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
DCUN1D3 Polyclonal antibody specifically detects DCUN1D3 in Human samples. It is validated for Immunohistochemistry.Specifications
DCUN1D3 | |
Polyclonal | |
Unconjugated | |
DCUN1D3 | |
1700020A13Rik; 44M2.4; BC030335; DCN1, defective in cullin neddylation 1, domain containing 3; DCN1, defective in cullin neddylation 1, domain containing 3 (S. cerevisiae); DCN1-like protein 3; DCN1-like protein 3-like; DCUN1 domain-containing protein 3; DCUN1D3; defective in cullin neddylation 1 domain containing 3; defective in cullin neddylation protein 1-like protein 3; hypothetical protein MGC48972; RGD1308893; SCCRO3; Squamous cell carcinoma-related oncogene 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
123879 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IWE4 | |
DCUN1D3 | |
Recombinant protein corresponding to Human DCUN1D3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only