missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDR2 [N456S] Recombinant Human Protein

Human DDR2, C-terminal fragment, amino acids R422-E855 (as in NCBI/Protein entry NP_006173.2), with a N456S mutation, activated, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.

492.00€ - 7490.00€

Specifications

For Use With (Application) Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
Formulation 50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol
Molecular Weight (g/mol) 77.8kDa
Storage Requirements Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Gene Alias MIG20a, NTRKR3, TKT, TYRO10
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15498833
View Documents Promotion Details
Invitrogen™
PV6172
10μg
492.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 09-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15408843
View Documents Promotion Details
Invitrogen™
PV6173
100μg
1678.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 09-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15418843
View Documents Promotion Details
Invitrogen™
PV6174
1mg
7490.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 09-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
77.8kDa
MIG20a, NTRKR3, TKT, TYRO10
Human
GST Tag, His Tag
50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
aa 422-855
Recombinant
Videos
SDS
Documents

Documents

Product Certifications