missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palintest™ De-Ion Pack, Photometer 7100
Click to view available options
Quantity:
10/Pk.
Unit Size:
Each
Description
- For use with the entry-level, multiparameter Photometer 7100.
- Makes up to 5 liters of deionized water.
Specifications
Specifications
| For Use With (Equipment) | Photometer 7100 |
| Quantity | 10/Pk. |
| Sufficient For | 5 L deionized water |
| Type | Water deionizing pack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction