Learn More
Trajan Scientific and Medical™ Deactivated (Methyl) Fused Silica Tubing
Product Code. 12746248
Click to view available options
Overall Length (Metric):
10 m
2 m
25 m
5 m
Diameter (Metric) Inner:
0.05 mm
0.11 mm
0.125 mm
0.25 mm
25 μm
60 μm
75 μm
100 μm
110 μm
150 μm
170 μm
220 μm
250 μm
320 μm
530 μm
Diameter (Metric) Outer:
0.31 mm
0.363 mm
170 μm
220 μm
285 μm
300 μm
363 μm
430 μm
680 μm
Includes:
1/Pk.
5/Pk.
Unit Size:
10m
25m
2m
5m
Each
Product Code. 12746248
Brand: Trajan Scientific and Medical™ 0624455
Only null left
Add to basket
Add to basket
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
  • Chemically inert and thermally stable up to 380°C
  • Suitable for organic and aqueous solvents
  • Ideal for biotechnology applications
Specifications
For Use With Organic and Aqueous Solvents
Type Tubing
Includes 1/Pk.
Diameter (Metric) Inner 100 μm
Diameter (Metric) Outer 363 μm
Overall Length (Metric) 25 m
Max. Operating Temperature (Metric) Thermally stable up to 380°C