Learn More
Borer Chemie™ deconex 22 PF
Product Code. p-7227859
Click to view available options
Quantity:
6 x 1.5 kg
1 x 10 kg
Unit Size:
10kg
9kg
Product Code. 17465102
Brand: Borer Chemie™ 501500.00D15P
Universal powder cleaner for laboratory glassware and utensils. The ecological solution deconex 22 PF shows a very high cleaning performance without chlorine, no surfactants and no phosphates. An acid rinse with a deconex cleaner is recommended.
  • Powder cleaner for automated cleaning
  • Ecological, phosphate-free solution
  • Perfect results already with 3-5 g/L
  • Can be rinsed off without leaving residues
  • For consistent and flawless results
  • Wide cleaning spectrum
  • Without chlorine, EDTA, NTA and surfactants
  • Acid rinse with deconex cleaner recommended
  • Technical support by the manufacturer
Specifications
Form Powder
pH ∼12.1 (1% Solution)
Composition Alkalis, complexing agents, corrosion inhibitor
Concentration Concentrate
Container Type Can
Content And Storage 5°C to 25°C
Quantity 6 x 1.5 kg
Weight (English) 19.8 lb.
Weight (Metric) 9 kg