missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decyl beta-D-maltopyranoside, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
1 g
5 g
Description
Decyl beta-D-maltopyranoside is used in a study to assess α-helical membrane protein crystallization. It has also been used in a study to investigate enantiomer separation of drugs by micellar electrokinetic chromatography. It aids in solubilizing the active cytochrome oxidase from mitochondria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Decyl beta-D-maltopyranoside is used in a study to assess α-helical membrane protein crystallization. It has also been used in a study to investigate enantiomer separation of drugs by micellar electrokinetic chromatography. It aids in solubilizing the active cytochrome oxidase from mitochondria.
Solubility
Soluble in methanol at 50mg/ml
Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Decyl beta-D-maltopyranoside is used in a study to assess α-helical membrane protein crystallization. It has also been used in a study to investigate enantiomer separation of drugs by micellar electrokinetic chromatography. It aids in solubilizing the active cytochrome oxidase from mitochondria.
Solubility
Soluble in methanol at 50mg/ml
Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Color | White |
Quantity | 500 mg |
Beilstein | 4923334 |
Solubility Information | Soluble in methanol at 50mg/ml |
Formula Weight | 482.56 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Decyl beta-D-maltopyranoside |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'