Learn More
Helios Italquartz Deep and Shallow Crucibles, Transparent
Product Code. 15868044
Click to view available options
Capacity (Metric):
15 cc
20 cc
5 cc
50 cc
Diameter (Metric) Outer:
29 mm
38 mm
41 mm
57 mm
Height (Metric):
19 mm
25 mm
37 mm
38 mm
Unit Size:
Each
Product Code. 15868044
Brand: Helios Italquartz 63P00005
Only null left
Add to basket
Add to basket
Deep and shallow crucibles constructed of transparent, durable, resistant, and chemically inert quartz glass.
  • Transparent
  • Deep and shallow forms available
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Color Transparent
Material Quartz
Capacity (Metric) 15 cc
Diameter (Metric) Outer 41 mm
Height (Metric) 25 mm
Product Type Crucible