missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Deerfit Lux Gloves
Protect your hands from the cold. Honeywell Safety Products™ are best for light and medium handling in cold conditions.
Brand: Honeywell Safety Products™ 2001615-08
Additional Details : Weight : 0.09900kg
Description
- Leather glove-fine black suede deerskin.
- Grip improved by buffed suede grain.
- Fully lined with Thinsulate™ for cold protection.
- Breathable, flexible, ultra lightweight, does not absorb humidity.
- Elasticized wrist for support and reduced thermal exchange.
- Black color makes the glove more soil proof.
Specifications
Dexterity: 4 | |
Buffed suede | |
Army-Defense, Energy, Electricity, Homeland Defense, Logistics, Service, Telecom, Transportation and Utility industries | |
Leather, Thinsulate™ | |
Cold | |
Black | |
EC Category PPE 1, EC Certificate Number: Autocertification, EC Attestation: EC Attestation |
Gunn | |
Elastic strip | |
Yes | |
Thinsulate™ | |
8 | |
25 cm |