Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale (EU)
Product Code. 17595825 Shop All Ohaus Products
Click to view available options
Capacity:
15 kg
30 kg
60 kg
150 kg
300 kg
600 kg
Model:
D33XW150B1L2
D33XW150B1L5
D33XW150B1X2
D33XW150B1X5
D33XW15B1R1
D33XW15B1R5
D33XW300B1V3
D33XW300B1X2
D33XW300B1X5
D33XW30B1R1
D33XW30B1R5
D33XW600B1V3
D33XW60B1L2
D33XW60B1L5
D33XW60B1R1
D33XW60B1R5
Readability:
2 g
5 g
10 g
20 g
50 g
100 g
Dimensions (L x W x H):
489 x 305 x 569 mm
646 x 305 x 122 mm
686 x 420 x 922 mm
787 x 500 x 922 mm
842 x 420 x 134 mm
941 x 500 x 150 mm
935 x 600 x 1222 mm
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17595825
Brand: OHAUS™ 30685247
Regulatory: This product is under eco-contribution tax.
Bench Scales for Basic Industrial Applications
  • Results are clearly shown using the large, bright backlit LCD display.
  • In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
  • This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
  • Indicator features a stainless steel housing and mounting bracket.
  • Features software for Parts Counting, Checkweighing and Totalization.
  • Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 100 Hours with Rechargeable Battery
Battery Type AC Adapter (Included);Rechargeable Battery (Included)
Connection Type Standard RS232 and optional USB Device or Ethernet
Construction Stainless steel indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet
Capacity 60 kg
Display Type 7-segment Multi-color Backlit LCD
Dimensions (LxW) Platform 550 x 420 mm
Readability 10 g
Model D33XW60B1L2
Temperature (Metric) Operating -10°C to +40°C, 85%RH, non-condensing
Weighing Modes Weighing, Checkweighing, Counting, Totalization.
Dimensions (L x W x H) 686 x 420 x 922 mm
Weight (Metric) 18.1 kg