Learn More
OHAUS™ Defender™ 5000 - D52 Bench Scale, Models TD52P QL
Product Code. 15978445 Shop All Ohaus Products
Click to view available options
Model:
D52P15RQDL2
D52P15RQDL2-M
D52P30RQDL2
D52P30RQDL2-M
D52P60RQDL2
D52P60RQDL2-M
Unit Size:
1 unit
Regulatory: This product is under eco-contribution tax.
Product Code. 15978445
Brand: OHAUS™ 30424315
Regulatory: This product is under eco-contribution tax.
OHAUS™ Defender™ 5000 Series multifunctional bench scale is ideal for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
  • User-friendly model with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
  • Durable construction with a stainless steel platform, sand gray powder-coated steel frame and aluminum load cell.
  • Defender meets NTEP/Measurement Canada or OIML/EC type approvals (depending on region).
  • Features common industrial modes and supports a 50000-item library with 1000 user profiles for easy data management.
  • Includes GMP/GLP with time/date for result traceability.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 20 Hours with Rechargeable Battery
Connection Type Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
Construction ABS plastic indicator, 304-grade stainless steel platform with sand gray powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet
Capacity 15 kg, 30 kg
Display Type Backlit LCD
Readability 1.000000 g, 2.000000 g
Model D52P30RQDL2
Temperature (Metric) Operating -10°C to +40°C
Weighing Modes Custom, Gram, Kilogram, Metric Tonnes, Ounce, Pound, Pound:Ounce, Tonne
Dimensions (L x W x H) 570 x 400 x 903 mm
Weight (Metric) 16.900 kg