Learn More
delta-Hexanolactone, 98%
Product Code. p-7024501
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11459536
Brand: Thermo Scientific Alfa Aesar A19274.14
Request Bulk or Custom Format
CAS: 823-22-3 | C6H10O2 | 114.144 g/mol
d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 823-22-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00083574
InChI Key RZTOWFMDBDPERY-UHFFFAOYSA-N
Synonym delta-hexalactone, delta-hexanolactone, delta-caprolactone, 5-methyl-delta-valerolactone, 5-hydroxyhexanoic acid lactone, tetrahydro-6-methyl-2h-pyran-2-one, 2h-pyran-2-one, tetrahydro-6-methyl, .delta.-caprolactone, 6-methylvalerolactone, hexanolactone
PubChem CID 13204
IUPAC Name 6-methyloxan-2-one
SMILES CC1CCCC(=O)O1
Specifications
Melting Point 18°C
Density 1.04
Boiling Point 231°C
Flash Point 105°C (221°F)
Refractive Index 1.451
Quantity 25 g
Beilstein 80501
Solubility Information Immiscible with water.
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material delta-Hexanolactone