Learn More
Honeywell™ Deltasafe Flame-Retardant M1 Polypropylene Coveralls with Hood
Catalog Number 11802991
missing translation for 'orderingAttributeHoverText'
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Deltasafe Polypropylene Coveralls are breathable, flame-retardant, single-use, non-ventilated protective suits designed to protect against particle contamination and chemical splash. Made of SMMS white polypropylene (60 g/m2) with an elasticated hood, ankles, and wrists.
  • Comfortable with Breathability Equal to a T-Shirt
  • Flame-Retardant and Antistatic Certified (EN 533 and EN 1149-1)
  • M1 and Radioactive Particle Certified (NF P92-507 and EN 1073-2)
  • Protection Level: General Requirements, Type 5 and Type 6 (EN 340, EN 13982-1, and EN 13034), EC Category PPE: 3
  • Recommended Industries: Chemical, Oil and Gas, Nuclear
  • Hazards: Airborne Particulate Contamination, Chemical Splash
TRUSTED_SUSTAINABILITY
Size Medium
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Hood Type Elastic
Sterility Non-sterile
Protection Class Protection type: 5, 6
Resistance Anti-static, Chemicals, Flame, Particle
Certifications/Compliance (Europe) EC Category PPE: 3, EC Certificate: Z2379/0563500/05, EC Attestation
Weight (Metric) 60 g/m2