Learn More
Honeywell™ Deltasafe Protective Jumpsuit
Breathable, flame retardant, single-use, non-ventilated jumpsuit for protection against particle contamination and chemical splashes.
Brand:  Honeywell™ 4571450-M
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11802991
  • 303.00€ / Pack of 25
Estimated Shipment
04-08-2021
Add to basket
Description
Description
  • Fire-retardant and M1-certified
  • Developed especially to avoid any risk when manipulating flammable substances
  • Antistatic-certified to avoid accumulation of electrostatic charges on the garment that may cause a risk of explosion or fire
  • Comfortable and breathable
Specifications
Specifications
Polypropylene
EC Category PPE: 3, EC Certificate: Z2379/0563500/05, EC Attestation
Unisex
60 g/m2
Protection type: 5, 6
White
Medium