Learn More
Density Standards (80 °C), Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C to 150°C
Brand: Paragon Scientific DEN80-02
Additional Details : CAS : 8042-47-5 Weight : 0.06000kg
Description
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols.
- Supplied in tamper evident packaging with a 2 year shelf life.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- Manufactured in the United Kingdom
Specifications
60 mL | |
0.8227 g/mL (80°C) | |
Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation |
Safety and Handling
- Density Standard (80 °C)
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : white mineral oil
Nominal values quoted