missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Desiccant Pan

The stainless steel Desiccant Pan holds granular desiccant (desiccant not included).

Brand:  Labconco™ 5532100

Additional Details : Weight : 0.50000kg

Product Code. 15309916

  • 117.00€ / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

  • Made from stainless steel
Specifications

Specifications

Desiccant Tray
Vacuum Dessicators
0.88 in.
0.5 kg
11 in.
28 cm
Stainless Steel
2.2 cm
1 lb.
28 cm
11 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions