missing translation for 'onlineSavingsMsg'
Learn More

Desmethylfluvoxamine, 1mg/ml in Acetonitrile, Lipomed

Product Code. 30457783
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30457783

Brand: Lipomed LPMFLV1889FB1LA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Desmethylfluvoxamine, 1mg/ml in Acetonitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethyl Fluvoxamine
CAS 192876-02-1
Analyte or Component Names Desmethyl Fluvoxamine
Shipping Condition Room Temperature
Molecular Formula C14 H19 F3 N2 O2
Concentration 1 mg/mL
Synonym (1 E)-5-hydroxy-1-[4-(trifluoromethyl)phenyl]-1-Pentanone O-(2-aminoethyl)oxime, 1-Pentanone, 5-hydroxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (E)-, Fluvoxamino acid, Fluvoxamine Maleate Imp. G (EP)
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES NCCO\N=C(/CCCCO)\c1ccc(cc1)C(F)(F)F
IUPAC Name (5 E)-5-(2-aminoethoxyimino)-5-[4-(trifluoromethyl)phenyl]pentan-1-ol
Solvent or Matrix Acetonitrile
Industry Type Pharmaceutical
Molecular Weight (g/mol) 304.31
Quantity 1 mL
Formula Weight 304.1399 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.