Learn More
Brady™ Destructible Tamper-evident Matte Vinyl Labels
Identify product tampering and prevent one-piece label removal with the B-351 tamper-resistant vinyl label material. Easily print almost anywhere with a Brady™ portable label printer.
Brand: Brady™ 170857
Description
- For use in asset tracking, rating plates, product security and tamper identification
- Designed to fracture to show signs of product tampering and prevent one-piece removal
- Made with a permanent adhesive that bonds tightly to almost any surface
Specifications
| Not Autoclavable | |
| Permanent Acrylic | |
| White | |
| Die-cut | |
| Top | |
| Label | |
| Asset tracking;Security identification | |
| 1 | |
| M5-20-351 | |
| 160 | |
| -40°F to 176°F | |
| 0.0028 in | |
| 25.40 mm (W) x 50.80 mm (H) | |
| 1 in | |
| 25.40 mm | |
| 0.5 in. | |
| M5-20-351 |
| 2011/65/EU, 2015/863 | |
| None | |
| Matte | |
| Vinyl | |
| Thermal Transfer | |
| Non-sterile | |
| BMP51;BMP53;M511;M510 | |
| -40°C | |
| 160 Label(s) / Box | |
| 25.40 mm (W) x 50.80 mm (H) | |
| -40°C to 80°C | |
| 0.071 mm | |
| 2 in. | |
| 25.40 mm | |
| 2 in | |
| 50.80 mm |