Learn More
Smeg™ Deterliquid™ Liquid Cleaner
Clean a wide range of glassware with this alkaline detergent. Smeg™ Deterliquid™ Liquid Cleaners achieve excellent washing results and optimize the thermal disinfection of laboratory instruments.
Brand: Smeg™ DET.RLQSP
Additional Details : Weight : 7.00000kg Transport : UN number : 1814 Chem class : 8 Pack group : II
Description
- For a variety of applications
- Available in 5 liter container
Specifications
SP Liquid Alkaline Deterliquid | |
Tin | |
1 Ea. | |
5 L |
Safety and Handling
- DETERLIQUID SP2
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P313-Get medical advice/attention.