Learn More
Smeg™ Deterliquid™ Liquid Cleaner
Clean a wide range of glassware with this alkaline detergent. Smeg™ Deterliquid™ Liquid Cleaners achieve excellent washing results and optimize the thermal disinfection of laboratory instruments.
Brand: Smeg™ DETERLQD2
Additional Details : CAS : 143239-08-1 Weight : 5.65000kg Transport : UN number : 1719 Chem class : 8 Pack group : II
Description
- For a variety of applications
- Available in 5 liter container
Specifications
D2 Liquid Alkaline Deterliquid | |
Canister | |
1 Ea. | |
5 L |
Safety and Handling
- DETERLIQUID D2
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Skin sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
- EUH206-Warning
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P302+P352-IF ON SKIN: Wash with plenty of water/.