Learn More
Fisherbrand™ Device Cover for Elmasonic X-tra ST Ultrasonic Cleaners
Provides a secure closure for ultrasonic cleaning baths.
Brand: Fisherbrand™
Additional Details : Weight : 1.00000kg
Description
- Flat design with handle
- Available in a range of sizes for all X-tra ST models
- Manufactured from stainless steel
- Accessory item
Specifications
Lid | |
Closure for X-tra ST Ultrasonic Cleaners | |
Flat |
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H | |
Handle |