Learn More
Taylor Wharton™ LD Series Cryogenic Storage Dewar
Product Code. p-8015007
Click to view available options
Capacity (Metric):
4 L
25 L
35 L
Unit Size:
Each
Product Code. 11778519
Brand: Taylor Wharton™ 25LDB
Only null left
Add to basket
Edge
Add to basket
With sealed vacuum space between the inner and outer shells to provide superior performance and insulation provides maximum holding times.
Specifications
Type Cryogenic Storage Dewar
Capacity (English) 6.6 gal.
Capacity (Metric) 25 L
Static Holding Time 109 Days