missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexamethasone-17-ketone (9alpha-Fluoro-11beta-hydroxy-16alpha-methylandrosta-1,4-diene-3,17-dione), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Dexamethasone-17-ketone (9alpha-Fluoro-11beta-hydroxy-16alpha-methylandrosta-1,4-diene-3,17-dione)
Specifications
Specifications
| Chemical Name or Material | 17-Oxo Dexamethasone |
| CAS | 1880-61-1 |
| Analyte or Component Names | 17-Oxo Dexamethasone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 F O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | NSC 50909, (11beta,16alpha)-9-Fluoro-11-hydroxy-16-methylandrosta-1,4-diene-3,17-dione, Androsta-1,4-diene-3,17-dione, 9-fluoro-11beta-hydroxy-16alpha-methyl- (7CI,8CI), 9-Fluoro-11beta-hydroxy-16alpha-methylandrosta-1,4-diene-3,17-dione, 9alpha-Fluoro-11beta-hydroxy-16alpha-methylandrosta-1,4-diene-3,17-dione |
| InChI Formula | InChI=1S/C20H25FO3/c1-11-8-15-14-5-4-12-9-13(22)6-7-19(12,3)20(14,21)16(23)10-18(15,2)17(11)24/h6-7,9,11,14-16,23H,4-5,8,10H2,1-3H3/t11-,14+,15+,16+,18+,19+,20+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)C1=O |
| IUPAC Name | (8S,9R,10S,11S,13S,14S,16R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,17-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction