Learn More
DiscoverX Dexamethasone
Product Code. 17372459
Encompass
Click to view available options
:
10 mg
Product Code. 17372459
Brand: DiscoverX 921132
Dexamethasone is an agonist for glucocorticoid receptor (GR). Provided as 10mg of dry powder.
Dexamethasone is an anti-inflammatory and immunosuppressant corticosteroid medication. Its glucocorticoid effect is 25 times more potent than cortisol. Uses include inflammation relief and treatment of arthritis, rheumatologic problems, skin diseases, allergies, asthma, chronic obstructive lung disease, and certain cancers. Acts as an agonist for glutocorticoid receptor (GR). Provided as 10mg of dry powder.
Specifications
Quantity 10 mg