Learn More
DiscoverX Dexamethasone
Click to view available options
:
10 mg
Description
Dexamethasone is an anti-inflammatory and immunosuppressant corticosteroid medication. Its glucocorticoid effect is 25 times more potent than cortisol. Uses include inflammation relief and treatment of arthritis, rheumatologic problems, skin diseases, allergies, asthma, chronic obstructive lung disease, and certain cancers. Acts as an agonist for glutocorticoid receptor (GR). Provided as 10mg of dry powder.
Specifications
Specifications
| Quantity | 10 mg |