Learn More
Dextran, MW ca 6,000, Thermo Scientific™
Dextran, also known as manninotriose, CAS # 9004-54-0, is a glucose polymer of bacterial origin.
Brand: Thermo Scientific Alfa Aesar J62775.22
197.70 EUR valid until 2026-09-25
BEST PRICE on Promo! Use promo code "27966" to get your promotional price.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Dextran is a complex branched polysaccharide of bacterial origin
• It primarily contains alpha-D C-1 – C-6 glycosidic linkages
• Alternate linkages account for the complex branching of dextran
Application
• Some research suggests that dextran can be used a carrier that helps target delivery of compounds to specific cells
• It is intended for protein isolation and centrifugation protocols, as well as other research applications