missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DFHBI
GREENER_CHOICE

Product Code. 18791688
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18791688

Brand: Tocris Bioscience™ 5609/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mimic of GFP fluorophore for imaging RNA in living cells

DFHBI is a mimic of green fluorescent protein (GFP) fluorophore for imaging RNA in living cells. DFHBI fluorescence is activated by binding to Spinach2 or Broccoli aptamers. Exhibits peak excitation maxima of 447 nm and peak fluorescence emission of 501 nm when bound to Spinach2.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5Z)-5-[(3,5-Difluoro-4-hydroxyphenyl)methylene]-3,5-dihydro-2,3-dimethyl-4H-imidazol-4-one
CAS 1241390-29-3
Quantity 10 mg
Target Light-up Aptamers
Molecular Formula C12H10F2N2O2
Formula Weight 252.22
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.