missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DFHBI

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
DFHBI is a mimic of green fluorescent protein (GFP) fluorophore for imaging RNA in living cells. DFHBI fluorescence is activated by binding to Spinach2 or Broccoli aptamers. Exhibits peak excitation maxima of 447 nm and peak fluorescence emission of 501 nm when bound to Spinach2.
Specifications
Specifications
| Chemical Name or Material | (5Z)-5-[(3,5-Difluoro-4-hydroxyphenyl)methylene]-3,5-dihydro-2,3-dimethyl-4H-imidazol-4-one |
| CAS | 1241390-29-3 |
| Quantity | 10 mg |
| Target | Light-up Aptamers |
| Molecular Formula | C12H10F2N2O2 |
| Formula Weight | 252.22 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction