missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ dI Phosphoramidite, standard grade, serum vial bottle

Product Code. p-4526896 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 μmol
250 mg
Unit Size:
100µmol
250mg
This item is not returnable. View return policy

Product Code. 17957102

Brand: Thermo Scientific™ 27174403

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

dI CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyinosine (dI) for use in oligonucleotide synthesis.

dI CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyinosine (dI) for use in oligonucleotide synthesis. The problem of degeneracy of the genetic code can be assuaged by utilizing universal bases, such as deoxyinosine (dI), in an oligonucleotide. Its unique structure allows for it to pair with all four bases in various ‘wobble’ structures, with an overall preferential order of base-pairing: dI-dC > dI-dA > dI-dG = dI-dT. This product is supplied in a 5 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyinosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C40H47N6O7P
Formula weight: 754.8 g/mol
CAS number: 141684-35-7
HPLC Purity: ≥98.0%; M-11 Impurity ≤0.01%
31P NMR Purity: ≥98%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type dI CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Deoxyinosine (dI)
Shipping Condition Room Temperature
Purity 98%
Quantity 100 μmol
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type DNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.