Learn More
Diaion™ WA20, ion exchange resin, weakly basic porous type, 2.5 meq/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046918.A1
Additional Details : Weight : 1.00000kg
Description
Diaion™ WA20 is used for the removal of the strong mineral acids in standard waste water applications. They can also be applied in the special process separation such as the treatment of organic solvents. Also used in deacidification, solvent treatment, refining of gelatin, adsorption of aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
300-1180μm Beads | |
Weakly basic porous type, 2.5meq/mL on poly(styrene-divinylbenzene) | |
300 to 1180μm | |
Not miscible or difficult to mix in water. |
Diaion™ WA20, ion exchange resin | |
Amine-like | |
1 kg |
RUO – Research Use Only