Learn More
Diazinon, Fluka™
Shop All Honeywell Fluka Products
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
missing translation for 'unitSize'
100mg
Beschreibung
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Spezifikation
Spezifikation
| Chemical Name or Material | Diazinon |
| Packaging | Glass Bottle |
| CAS | 333-41-5 |
| Color | Colorless |
| Boiling Point | Decomposes below the boiling point. |
| Synonym | O,O-diethyl O-2-isopropyl-6-methylpyrimidin-4-yl phosphorothioate |
| Industry Type | Environmental Analysis |
| Molecular Weight (g/mol) | 304.3 |
| Quantity | 100 mg |
| Grade | Analytical Standard |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur