missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiethylsilane, 94%, Thermo Scientific Chemicals

Catalog Number p-7043875
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
1719-53-5
C4H10Cl2Si
157.11
MFCD00000493
BYLOHCRAPOSXLY-UHFFFAOYSA-N
diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl
15589
dichloro(diethyl)silane
CC[Si](Cl)(Cl)CC
This item is not returnable. View return policy
1719-53-5
C4H10Cl2Si
157.11
MFCD00000493
BYLOHCRAPOSXLY-UHFFFAOYSA-N
diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl
15589
dichloro(diethyl)silane
CC[Si](Cl)(Cl)CC

Dichlorodiethylsilane is used in ethchlorvynol assay. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorodiethylsilane is used in ethchlorvynol assay. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Slightly soluble in water and ether.

Notes
Moisture Sensitive. Store at 4°C. Protect from moisture.Incompatible materials are Oxidizing agents, Bases, Water.
TRUSTED_SUSTAINABILITY
Boiling Point 129°C to 131°C
Melting Point -97°C
Quantity 10 g
UN Number UN1767
Beilstein 605313
Flash Point 28°C (82°F)
Solubility Information Slightly soluble in water and ether.
Formula Weight 157.12
Percent Purity 94%
Odor Acrid
Refractive Index 1.431
Sensitivity Moisture sensitive
Density 1.055
Chemical Name or Material Dichlorodiethylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'