missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals

Product Code. 10351434 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10351434

Brand: Thermo Scientific Acros 310240500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676-83-5
Molecular Formula CH3Cl2P
Molecular Weight (g/mol) 116.909
MDL Number MFCD00013616
InChI Key CDPKWOKGVUHZFR-UHFFFAOYSA-N
Synonym methyldichlorophosphine, dichloromethylphosphine, methylphosphonous dichloride, dichloro methyl phosphane, phosphonous dichloride, p-methyl, ch3pcl2, phosphonous dichloride, methyl, methyl phosphonous dichloride, methylphosphorus dichloride, phosphine, dichloromethyl
PubChem CID 61194
IUPAC Name dichloro(methyl)phosphane
SMILES CP(Cl)Cl

Specifications

Density 1.3000g/mL
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula CH3PCl2
Quantity 50 g
Formula Weight 116.92
Grade Technical
Chemical Name or Material Dichloromethylphosphine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.