missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol, Thermo Scientific Chemicals

Catalog Number p-4510191
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
77-73-6, 98-29-3
C10H12
132.2
HECLRDQVFMWTQS-UHFFFAOYSA-N
dicyclopentadiene, biscyclopentadiene, cyclopentadiene dimer, bicyclopentadiene, dicyklopentadien, dimer cyklopentadienu, dcpd, 4,7-methano-1h-indene, 3a,4,7,7a-tetrahydro, 3a,4,7,7a-tetrahydro-1h-4,7-methanoindene, 1,3-cyclopentadiene, dimer
6492
CHEBI:34695
C1C=CC2C1C3CC2C=C3
This item is not returnable. View return policy
77-73-6, 98-29-3
C10H12
132.2
HECLRDQVFMWTQS-UHFFFAOYSA-N
dicyclopentadiene, biscyclopentadiene, cyclopentadiene dimer, bicyclopentadiene, dicyklopentadien, dimer cyklopentadienu, dcpd, 4,7-methano-1h-indene, 3a,4,7,7a-tetrahydro, 3a,4,7,7a-tetrahydro-1h-4,7-methanoindene, 1,3-cyclopentadiene, dimer
6492
CHEBI:34695
C1C=CC2C1C3CC2C=C3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 11.0°C to 13.0°C
Density 0.9820g/mL
Boiling Point 170.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (endo-Dicyclopentadiene) (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.5120
Quantity 100 mL
Beilstein 05,II,391
Merck Index 14,2739
Specific Gravity 0.982
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ethanol,diethyl ether
Stabilizer 100 to 200ppm p-tert.-butylcatechol
Formula Weight 132.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Dicyclopentadiene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dicyclopentadiene, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'