missing translation for 'onlineSavingsMsg'
Learn More

Diethyl diethylmalonate, 98%

Product Code. 11470268
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11470268

Brand: Thermo Scientific Alfa Aesar B23810.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-25-8 | C11H20O4 | 216.277 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-25-8
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00009132
InChI Key ZKBBUZRGPULIRN-UHFFFAOYSA-N
Synonym diethyl diethylmalonate, diethyl 2,2-diethylmalonate, diethylmalonic ester, propanedioic acid, diethyl-, diethyl ester, diethylmalonate diethyl ester, diethylmalonic acid diethyl ester, diethylmalonic acid, diethyl ester, unii-ceh13944yq, 1,3-diethyl 2,2-diethylpropanedioate, diethyl diethylmalonate diethyl
PubChem CID 66165
IUPAC Name diethyl 2,2-diethylpropanedioate
SMILES CCC(CC)(C(=O)OCC)C(=O)OCC

Specifications

Density 0.99
Boiling Point 228°C to 230°C
Flash Point 94°C (202°F)
Refractive Index 1.423
Quantity 25 g
Merck Index 14,3789
Formula Weight 216.28
Percent Purity 98%
Chemical Name or Material Diethyl diethylmalonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.